Learn More
Cayman Chemical -PraeruptrIn A 100mg
SDP
Catalog No. 502933826
Encompass_Preferred
A coumarin derivative; prevents LPS-induced increases in IL-1β and TNF-α at 25-100 µM and IL-6 at 25-100 µM; inhibits potassium chloride-induced contraction of isolated rabbit trachea by 50 and 100% at 4.8 and 30 µM, respectively; reduces edema in a croton oil mouse model of inflammation at 0.3 µmol/cm2 applied topically
Catalog No. 50-293-3826 Supplier Cayman Chemical Supplier No. 24915100 MG
A coumarin derivative; prevents LPS-induced increases in IL-1β and TNF-α at 25-100 µM and IL-6 at 25-100 µM; inhibits potassium chloride-induced contraction of isolated rabbit trachea by 50 and 100% at 4.8 and 30 µM, respectively; reduces edema in a croton oil mouse model of inflammation at 0.3 µmol/cm2 applied topically