Learn More
PUROLITE COMPANY Affinity chromatography resin, 5 ml MiniChrom Column, average particle size 70 μm, dynamic binding capacity 50 g/L, pH stability 2 - 13, alkaline stability 0.1 M NaOH
Catalog No. NC3525563
Encompass
Affinity chromatography resin, 5 ml MiniChrom Column, average particle size 70 μm, dynamic binding capacity 50 g/L, pH stability 2 - 13, alkaline stability 0.1 M NaOH
Catalog No. NC3525563 Supplier PUROLITE COMPANY Supplier No. PR00670175
Affinity chromatography resin, 5 ml MiniChrom Column, average particle size 70 μm, dynamic binding capacity 50 g/L, pH stability 2 - 13, alkaline stability 0.1 M NaOH