Learn More
PUROLITE COMPANY Praesto Jetted A50 HipH, Protein A affinity resin, 100 ml, 50 μm particle size, Elution pH 3 - 5, Dynamic Binding Capacity 60 g/L, Alkaline Stability 0.1 M NaOH for 120 h
Catalog No. NC3345922
Encompass
Praesto Jetted A50 HipH, Protein A affinity resin, 100 ml, 50 μm particle size, Elution pH 3 - 5, Dynamic Binding Capacity 60 g/L, Alkaline Stability 0.1 M NaOH for 120 h
Catalog No. NC3345922 Supplier PUROLITE COMPANY Supplier No. PR00530164
Praesto Jetted A50 HipH, Protein A affinity resin, 100 ml, 50 μm particle size, Elution pH 3 - 5, Dynamic Binding Capacity 60 g/L, Alkaline Stability 0.1 M NaOH for 120 h