Learn More
TARGETMOL CHEMICALS INC PRALATREXATE 10MG
SDP
Catalog No. 502576595
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pralatrexate (10-Propargyl-10-deazaaminopterin) is a folate analogue inhibitor of dihydrofolate reductase (DHFR) exhibiting high affinity for reduced folate carrier-1 (RFC-1) with antineoplastic and immunosuppressive activities. purity: 98%
Catalog No. 50-257-6595 Supplier TARGETMOL CHEMICALS INC Supplier No. T612010MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pralatrexate (10-Propargyl-10-deazaaminopterin) is a folate analogue inhibitor of dihydrofolate reductase (DHFR) exhibiting high affinity for reduced folate carrier-1 (RFC-1) with antineoplastic and immunosuppressive activities. purity: 98%