Learn More
Apexbio Technology LLC Pralatrexate, 146464-95-1, MFCD00920897, 10 mg
SDP
Supplier:  Apexbio Technology LLC A435010
Encompass_Preferred
MF: C23H23N7O5, MW: 477.47, Solubility: 23.85 mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. Pralatrexate is an inhibitor of DHFR with Ki value of 45 nM.
Catalog No. 50-101-1847