Learn More
Apexbio Technology LLC Pralatrexate 146464-95-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061613
Encompass_Preferred
Pralatrexate (CAS 146464-95-1) is a small molecule inhibitor targeting dihydrofolate reductase (DHFR) with a Ki value of 45 nM DHFR mediates the reduction of dihydrofolate to tetrahydrofolate a critical cofactor in purine thymidylate and amino acid biosynthesis Pralatrexate exhibits high affinity for folylpolyglutamate synthetase (FPGS) and reduced folate carrier-1 (RFC-1) facilitating intracellular accumulation It demonstrates notable antiproliferative activity against various human cancer cell lines in vitro and reduces tumor growth in NSCLC xenograft models indicating its suitability for oncology research
Catalog No. 50-000-61613 Supplier Apexbio Technology LLC Supplier No. A43505.1
Pralatrexate (CAS 146464-95-1) is a small molecule inhibitor targeting dihydrofolate reductase (DHFR) with a Ki value of 45 nM DHFR mediates the reduction of dihydrofolate to tetrahydrofolate a critical cofactor in purine thymidylate and amino acid biosynthesis Pralatrexate exhibits high affinity for folylpolyglutamate synthetase (FPGS) and reduced folate carrier-1 (RFC-1) facilitating intracellular accumulation It demonstrates notable antiproliferative activity against various human cancer cell lines in vitro and reduces tumor growth in NSCLC xenograft models indicating its suitability for oncology research