Learn More
Medchemexpress LLC Praliciguat | 1628730-49-3 | 99.35% | 534.36 | 100 MG
SDP
Catalog No. 5000345131
Encompass_Preferred
Praliciguat is a potent and orally active soluble guanylate cyclase (sGC) activator. It increases cGMP via the nitric oxide (NO)-sGC pathway, inhibits the expression of proinflammatory cytokines and apoptosis, and promotes vasodilation. It is used for research on metabolic and cardiovascular diseases, such as hypertension, diabetes, and heart failure.
  • Potent and orally active soluble guanylate cyclase (sGC) activator
  • Increases cGMP via the nitric oxide (NO)-sGC pathway
  • Inhibits the expression of proinflammatory cytokines
  • Inhibits apoptosis
  • Promotes vasodilation
  • Stimulates soluble guanylate cyclase in HEK-293 cells with an EC50 of 197 nM
  • Reduces arterial pressure (MAP) in normotensive and hypertensive rats
  • Attenuates proteinuria in obese ZSF1 rats model of diabetic nephropathy
  • Attenuates inflammation, fibrosis, and end-organ damage in the Dahl rats model of cardiorenal failure
Catalog No. 50-003-45131 Supplier Medchemexpress LLC Supplier No. HY10903950MG
Praliciguat is a potent and orally active soluble guanylate cyclase (sGC) activator. It increases cGMP via the nitric oxide (NO)-sGC pathway, inhibits the expression of proinflammatory cytokines and apoptosis, and promotes vasodilation. It is used for research on metabolic and cardiovascular diseases, such as hypertension, diabetes, and heart failure.
  • Potent and orally active soluble guanylate cyclase (sGC) activator
  • Increases cGMP via the nitric oxide (NO)-sGC pathway
  • Inhibits the expression of proinflammatory cytokines
  • Inhibits apoptosis
  • Promotes vasodilation
  • Stimulates soluble guanylate cyclase in HEK-293 cells with an EC50 of 197 nM
  • Reduces arterial pressure (MAP) in normotensive and hypertensive rats
  • Attenuates proteinuria in obese ZSF1 rats model of diabetic nephropathy
  • Attenuates inflammation, fibrosis, and end-organ damage in the Dahl rats model of cardiorenal failure