Learn More
Medchemexpress LLC Praliciguat | 1628730-49-3 | 99.35% | 534.36 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY109039100MG
Encompass_Preferred
Praliciguat (IW-1973) is a potent and orally active soluble guanylate cyclase (sGC) activator. It increases cGMP via the nitric oxide (NO)-sGC pathway, inhibits the expression of proinflammatory cytokines and apoptosis, and promotes vasodilation. This compound is used for research into metabolic and cardiovascular diseases, including hypertension, diabetes, and heart failure.
  • Potent and orally active sGC activator
  • Increases cGMP via the NO-sGC pathway
  • Inhibits proinflammatory cytokines and apoptosis
  • Promotes vasodilation
  • Used for research into metabolic and cardiovascular diseases
  • Suitable for studying hypertension, diabetes, and heart failure
Catalog No. 50-003-46142