Learn More
Medchemexpress LLC Praliciguat | 1628730-49-3 | 99.4% | 534.36 | 1 ML
SDP
Catalog No. 5000344598
Encompass_Preferred
Praliciguat (IW-1973) is a potent and orally active soluble guanylate cyclase (sGC) activator. It increases cGMP through the nitric oxide (NO)-sGC pathway. Praliciguat can also inhibit the expression of proinflammatory cytokines and apoptosis, and it promotes vasodilation. This compound can be used in research for metabolic diseases and cardiovascular diseases, including hypertension, diabetes, and heart failure.
  • Potent and orally active soluble guanylate cyclase (sGC) activator.
  • Increases cGMP via the nitric oxide (NO)-sGC pathway.
  • Inhibits the expression of proinflammatory cytokines.
  • Inhibits apoptosis.
  • Promotes vasodilation.
  • Useful for research in metabolic diseases and cardiovascular diseases, such as hypertension, diabetes, and heart failure.
Catalog No. 50-003-44598 Supplier Medchemexpress LLC Supplier No. HY10903910MG
Praliciguat (IW-1973) is a potent and orally active soluble guanylate cyclase (sGC) activator. It increases cGMP through the nitric oxide (NO)-sGC pathway. Praliciguat can also inhibit the expression of proinflammatory cytokines and apoptosis, and it promotes vasodilation. This compound can be used in research for metabolic diseases and cardiovascular diseases, including hypertension, diabetes, and heart failure.
  • Potent and orally active soluble guanylate cyclase (sGC) activator.
  • Increases cGMP via the nitric oxide (NO)-sGC pathway.
  • Inhibits the expression of proinflammatory cytokines.
  • Inhibits apoptosis.
  • Promotes vasodilation.
  • Useful for research in metabolic diseases and cardiovascular diseases, such as hypertension, diabetes, and heart failure.