Learn More
Apexbio Technology LLC PRALIDOXIME-CHLORIDE-200MG
SDP
Catalog No. 5000569371
Encompass_Preferred
Pralidoxime chloride (CAS No 51-15-0) is widely studied in biomedical research as a cholinesterase reactivator particularly in the context of organophosphate poisoning Originating from the need to counteract nerve agents its mechanism of action involves binding to the phosphorylated active site of acetylcholinesterase thereby restoring its function Commonly its in vitro activity is observed in the nanomolar to low micromolar range allowing for precise modulation of enzyme activity It is frequently utilized in cell models and animal studies significantly contributing to drug screening processes aimed at evaluating antidotal efficacy This compound enables researchers to test and develop new treatments for exposure to toxic organophosphates and its use in experiments often involves variable concentrations tailored to the specific research design and goals
Catalog No. 50-005-69371 Supplier Apexbio Technology LLC Supplier No. C7193200
Pralidoxime chloride (CAS No 51-15-0) is widely studied in biomedical research as a cholinesterase reactivator particularly in the context of organophosphate poisoning Originating from the need to counteract nerve agents its mechanism of action involves binding to the phosphorylated active site of acetylcholinesterase thereby restoring its function Commonly its in vitro activity is observed in the nanomolar to low micromolar range allowing for precise modulation of enzyme activity It is frequently utilized in cell models and animal studies significantly contributing to drug screening processes aimed at evaluating antidotal efficacy This compound enables researchers to test and develop new treatments for exposure to toxic organophosphates and its use in experiments often involves variable concentrations tailored to the specific research design and goals