Learn More
Medchemexpress LLC Pralidoxime chloride | 51-15-0 | 99.8% | 1 ML
SDP
Catalog No. 5000413581
Encompass_Preferred
Pralidoxime chloride is a potent reactivator of acetylcholinesterase (AChE). It effectively reactivates nerve agent-inhibited AChE through a direct nucleophilic attack by its oxime moiety on the phosphorus center of the bound nerve agent. This makes Pralidoxime chloride a recognized antidote for organophosphate poisoning, crucial for counteracting the effects of such toxins.
  • Potent reactivator of acetylcholinesterase (AChE).
  • Reactivates nerve agent-inhibited AChE.
  • Antidote for organophosphate poisoning.
  • For research use only.
Catalog No. 50-004-13581 Supplier Medchemexpress LLC Supplier No. HYB120010MM/1ML
Pralidoxime chloride is a potent reactivator of acetylcholinesterase (AChE). It effectively reactivates nerve agent-inhibited AChE through a direct nucleophilic attack by its oxime moiety on the phosphorus center of the bound nerve agent. This makes Pralidoxime chloride a recognized antidote for organophosphate poisoning, crucial for counteracting the effects of such toxins.
  • Potent reactivator of acetylcholinesterase (AChE).
  • Reactivates nerve agent-inhibited AChE.
  • Antidote for organophosphate poisoning.
  • For research use only.