Learn More
Selleck Chemical LLC Pralidoxime Iodide S5100-25mg
SDP
Catalog No. 503112058
Encompass_Preferred
Pralidoxime Iodide (2-PAM) is an antidote approved for reactivation of inhibited acetylcholinesterase (AChE) in organophosphate poisoning
Catalog No. 50-311-2058 Supplier Selleck Chemical LLC Supplier No. S510025MG
Pralidoxime Iodide (2-PAM) is an antidote approved for reactivation of inhibited acetylcholinesterase (AChE) in organophosphate poisoning