Learn More
Medchemexpress LLC Pralidoxime iodide | 94-63-3 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB1738A10MM/1ML
Encompass_Preferred
Pralidoxime iodide is a potent reactivator of acetylcholinesterase (AChE) that works by direct nucleophilic attack of its oxime moiety on the phosphorus center of the bound nerve agent, making it an antidote for organophosphate poisoning.
  • Potent reactivator of acetylcholinesterase (AChE)
  • Reactivates nerve agent-inhibited AChE
  • Antidote for organophosphate poisoning
Catalog No. 50-004-12571