Learn More
Medchemexpress LLC Pralidoxime (iodide) | 94-63-3 | 98.0% | 250 MG
SDP
Catalog No. 5000414490
Encompass_Preferred
Pralidoxime iodide is a potent reactivator of acetylcholinesterase (AChE). It reactivates nerve agent-inhibited AChE through direct nucleophilic attack by the oxime moiety on the phosphorus center of the bound nerve agent. It is also an antidote for organophosphate poisoning.
  • Potent reactivator of acetylcholinesterase (AChE).
  • Reactivates nerve agent-inhibited AChE.
  • Antidote for organophosphate poisoning.
Catalog No. 50-004-14490 Supplier Medchemexpress LLC Supplier No. 250MGHYB1738A
Pralidoxime iodide is a potent reactivator of acetylcholinesterase (AChE). It reactivates nerve agent-inhibited AChE through direct nucleophilic attack by the oxime moiety on the phosphorus center of the bound nerve agent. It is also an antidote for organophosphate poisoning.
  • Potent reactivator of acetylcholinesterase (AChE).
  • Reactivates nerve agent-inhibited AChE.
  • Antidote for organophosphate poisoning.