Learn More
Medchemexpress LLC HY-19676 1mg , Pralnacasan CAS:192755-52-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY196761MG
Encompass_Preferred
HY-19676 1mg Pralnacasan CAS: 192755-52-5 Pralnacasan (VX-740) is a potent, selective, non-peptide and orally active interleukin-1β converting enzyme (ICE, caspase 1) inhibitor with a Ki of 1.4 nM. Pralnacasan inhibits proinflammatory cytokines IL-18, IL-1β , and IFN-γ. Pralnacasan has the potential for osteoarthritis and rheumatoid arthritis treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1510