Learn More
Medchemexpress LLC Pralnacasan (VX-740) | 192755-52-5 | 99.8% | 523.54 | 5 MG
SDP
Catalog No. 5000400687
Encompass_Preferred
Pralnacasan (VX-740) is a potent, selective, non-peptide and orally active interleukin-1β converting enzyme (ICE, caspase 1) inhibitor with a Ki of 1.4 nM. It inhibits proinflammatory cytokines IL-18, IL-1β , and IFN-γ. Pralnacasan has the potential for osteoarthritis and rheumatoid arthritis treatment.
  • Potent, selective, non-peptide and orally active interleukin-1β converting enzyme (ICE, caspase 1) inhibitor with a Ki of 1.4 nM.
  • Inhibits proinflammatory cytokines IL-18, IL-1β , and IFN-γ.
  • Potential for osteoarthritis and rheumatoid arthritis treatment.
  • Reduces joint damage in two murine models of osteoarthritis.
  • Does not appear to affect the weight of the animals in an animal model.
Catalog No. 50-004-00687 Supplier Medchemexpress LLC Supplier No. HY196765MG
Pralnacasan (VX-740) is a potent, selective, non-peptide and orally active interleukin-1β converting enzyme (ICE, caspase 1) inhibitor with a Ki of 1.4 nM. It inhibits proinflammatory cytokines IL-18, IL-1β , and IFN-γ. Pralnacasan has the potential for osteoarthritis and rheumatoid arthritis treatment.
  • Potent, selective, non-peptide and orally active interleukin-1β converting enzyme (ICE, caspase 1) inhibitor with a Ki of 1.4 nM.
  • Inhibits proinflammatory cytokines IL-18, IL-1β , and IFN-γ.
  • Potential for osteoarthritis and rheumatoid arthritis treatment.
  • Reduces joint damage in two murine models of osteoarthritis.
  • Does not appear to affect the weight of the animals in an animal model.