Learn More
Cayman Chemical Pralsetinib, Purity: ≥98%, CAS: 2097132-94-8, 10 mg
SDP
Catalog No. 502947406
Encompass_Preferred
An inhibitor of RET (IC50s = 0.4, 0.3, 0.4, and 0.4 nM for wild-type RET, RETV804L, RETV804M, and RETM918T, respectively); selective for RET over VEGFR2 (IC50 = 35 nM) and a panel of 371 kinases at 300 nM; inhibits a CCDC6-RET fusion protein and autophosphorylation of RET in Ba/F3 cells expressing a KIF5B-RET fusion protein (IC50s = 0.4 and 5 nM, respectively); induces tumor regression in a KIF5B-RET-positive PDX mouse model of NSCLC at 30 mg/kg
Catalog No. 50-294-7406 Supplier Cayman Chemical Supplier No. 3318110 MG
An inhibitor of RET (IC50s = 0.4, 0.3, 0.4, and 0.4 nM for wild-type RET, RETV804L, RETV804M, and RETM918T, respectively); selective for RET over VEGFR2 (IC50 = 35 nM) and a panel of 371 kinases at 300 nM; inhibits a CCDC6-RET fusion protein and autophosphorylation of RET in Ba/F3 cells expressing a KIF5B-RET fusion protein (IC50s = 0.4 and 5 nM, respectively); induces tumor regression in a KIF5B-RET-positive PDX mouse model of NSCLC at 30 mg/kg