Learn More
Medchemexpress LLC HY-112301 5mg Medchemexpress, Pralsetinib CAS:2097132-94-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1123015MG
Encompass_Preferred
Medchemexpress, HY-112301 5mg Pralsetinib CAS:2097132-94-8 Pralsetinib (BLU-667) is a highly potent, selective RET inhibitor. Pralsetinib (BLU-667) inhibits WT RET, RET mutants V804L, V804M, M918T and CCDC6-RET fusion with IC50s of 0.4, 0.3, 0.4, 0.4, and 0.4 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9076