Learn More
Pramana Pramana Pick Basket
Description
- Pramana Pick Basket for High Throughput System
- Robotic Handler
- 120 slide pick basket supporting 1x3 slides
- Compatible with Pramana High Throughput Systems with Robotic Handler.
Specifications
Specifications
| Product Type | Slide Basket |
| For Use With (Equipment) | High Throughput System with Robotic Handler |