Learn More
Biocare Medical, LLC Biocare Medical PRAME Monoclonal Antibody, Enhanced Diagnostic Accuracy, High Specificity, 0.5 mL
SDP
Supplier:  Biocare Medical, LLC ACI3252B
Encompass_Preferred
The Biocare Medical PRAME Monoclonal Antibody is a 0.5 mL concentrate designed for identifying PRAME protein in FFPE human tissues via immunohistochemistry. This product enhances diagnostic accuracy by differentiating benign melanocytic nevi from melanoma, and aids in assessing tumor margins. Its high specificity and optimized formulation make it invaluable for detecting various malignancies, positioning it as a superior choice in cancer diagnostics.
Catalog No. NC2089113