Learn More
Aviva Systems Biology Corp PRAME Antibody - N-terminal region (ARP55981_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55981_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRAME. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PRAME. Alternate names: MAPE, OIP4, CT130ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-61682