Learn More
Aviva Systems Biology Corp PRAMEF10 Antibody - middle region (ARP54493_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54493_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRAMEF10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRAMEF10. Alternate names: MGC138413, MGC138415, RP5-845O24.7ApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-014-61224