Learn More
Selleck Chemical LLC Pramipexole S2460-20mg
SDP
Catalog No. 503108859
Encompass_Preferred
Pramipexole (SND 919 Mirapexin Sifrol Mirapex) is a partial/full D2S D2L D3 D4 receptor agonist with a Ki of 3 9 2 2 0 5 and 5 1 nM for D2S D2L D3 D4 receptor respectively
Catalog No. 50-310-8859 Supplier Selleck Chemical LLC Supplier No. S246020MG
Pramipexole (SND 919 Mirapexin Sifrol Mirapex) is a partial/full D2S D2L D3 D4 receptor agonist with a Ki of 3 9 2 2 0 5 and 5 1 nM for D2S D2L D3 D4 receptor respectively