Learn More
Apexbio Technology LLC Pramipexole 2HCl Monohydrate 191217-81-9 50mg
SDP
Catalog No. 5000090507
Encompass_Preferred
Pramipexole 2HCl Monohydrate is a synthetic non-ergoline dopamine receptor agonist targeting dopamine receptor subtypes particularly D3 D2S D2L and D4 It is designed to activate these receptors thereby modulating dopaminergic transmission pathways Pramipexole 2HCl Monohydrate exerts its biological activity primarily through selective receptor activation Based on these pharmacological properties Pramipexole 2HCl Monohydrate holds research potential in studies of receptor-ligand interactions receptor subtype functional analysis and dopamine-related neurologic or psychiatric disorders including Parkinson s disease and restless legs syndrome
Catalog No. 50-000-90507 Supplier Apexbio Technology LLC Supplier No. B148750
Pramipexole 2HCl Monohydrate is a synthetic non-ergoline dopamine receptor agonist targeting dopamine receptor subtypes particularly D3 D2S D2L and D4 It is designed to activate these receptors thereby modulating dopaminergic transmission pathways Pramipexole 2HCl Monohydrate exerts its biological activity primarily through selective receptor activation Based on these pharmacological properties Pramipexole 2HCl Monohydrate holds research potential in studies of receptor-ligand interactions receptor subtype functional analysis and dopamine-related neurologic or psychiatric disorders including Parkinson s disease and restless legs syndrome