Learn More
Apexbio Technology LLC Pramipexole dihydrochloride 104632-25-9 50mg
SDP
Catalog No. 5000096787
Encompass_Preferred
Pramipexole dihydrochloride is a small-molecule agonist targeting dopamine receptors exhibiting selective affinity toward the D3 dopamine receptor subtype and lower affinity for the D2L D2S and D4 subtypes It is designed to stimulate dopamine receptor-mediated signaling thereby modulating dopaminergic pathways involved in motor function Pramipexole dihydrochloride exerts its biological activity primarily through dopamine receptor agonism and also demonstrates receptor-independent neuroprotective effects by attenuating mitochondrial reactive oxygen species (ROS) production and decreasing apoptosis-related signaling cascades Based on these pharmacological properties pramipexole dihydrochloride holds research potential in exploring dopaminergic system involvement and neuroprotective mechanisms relevant to neurodegenerative conditions such as Parkinson s disease and restless legs syndrome
Catalog No. 50-000-96787 Supplier Apexbio Technology LLC Supplier No. A373350
Pramipexole dihydrochloride is a small-molecule agonist targeting dopamine receptors exhibiting selective affinity toward the D3 dopamine receptor subtype and lower affinity for the D2L D2S and D4 subtypes It is designed to stimulate dopamine receptor-mediated signaling thereby modulating dopaminergic pathways involved in motor function Pramipexole dihydrochloride exerts its biological activity primarily through dopamine receptor agonism and also demonstrates receptor-independent neuroprotective effects by attenuating mitochondrial reactive oxygen species (ROS) production and decreasing apoptosis-related signaling cascades Based on these pharmacological properties pramipexole dihydrochloride holds research potential in exploring dopaminergic system involvement and neuroprotective mechanisms relevant to neurodegenerative conditions such as Parkinson s disease and restless legs syndrome