Learn More
Medchemexpress LLC Pramipexole dihydrochloride | 104632-25-9 | 99.7% | 284.25 | 1 ML
SDP
Catalog No. 5000384559
Encompass_Preferred
Pramipexole dihydrochloride is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist. It can be used for the research of Parkinson's disease (PD) and restless legs syndrome (RLS).
  • Selective dopamine D2-type receptor agonist
  • BBB penetrant
  • Useful for research into Parkinson's disease (PD) and restless legs syndrome (RLS)
  • Cytoprotective properties
  • Reduces infarction volume
  • Improves neurological recovery
  • Prevents ischemic cell death
Catalog No. 50-003-84559 Supplier Medchemexpress LLC Supplier No. HY1735510MM/1ML
Pramipexole dihydrochloride is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist. It can be used for the research of Parkinson's disease (PD) and restless legs syndrome (RLS).
  • Selective dopamine D2-type receptor agonist
  • BBB penetrant
  • Useful for research into Parkinson's disease (PD) and restless legs syndrome (RLS)
  • Cytoprotective properties
  • Reduces infarction volume
  • Improves neurological recovery
  • Prevents ischemic cell death