Learn More
Medchemexpress LLC Pramipexole dihydrochloride | 104632-25-9 | MFCD00876894 | 99.7% | 284.25 g/mol | C10H19Cl2N3S | 10 MG
SDP
Catalog No. 5000295014
Encompass_Preferred
Pramipexole dihydrochloride is a research-grade dopamine D2-type receptor agonist provided as a white to off-white solid salt. It is commonly used in neuroscience and pharmacology studies, including models of Parkinson's disease and restless legs syndrome, where selective D2 agonism and blood-brain barrier penetration are required.
  • High HPLC purity (99.73%).
  • White to off-white solid for consistent handling.
  • Suitable for in vitro and in vivo neuroscience research.
  • Blood-brain barrier penetrant dopamine D2-type receptor agonist.
  • Available in multiple small research pack sizes.
  • Stable when stored sealed at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-95014 Supplier Medchemexpress LLC Supplier No. HY1735510MG
Pramipexole dihydrochloride is a research-grade dopamine D2-type receptor agonist provided as a white to off-white solid salt. It is commonly used in neuroscience and pharmacology studies, including models of Parkinson's disease and restless legs syndrome, where selective D2 agonism and blood-brain barrier penetration are required.
  • High HPLC purity (99.73%).
  • White to off-white solid for consistent handling.
  • Suitable for in vitro and in vivo neuroscience research.
  • Blood-brain barrier penetrant dopamine D2-type receptor agonist.
  • Available in multiple small research pack sizes.
  • Stable when stored sealed at 4°C; in solvent: -80°C (6 months), -20°C (1 month).