Learn More
Selleck Chemical LLC Pramipexole dihydrochloride S5066-25mg
SDP
Catalog No. 503112222
Encompass_Preferred
Pramipexole dihydrochloride (SND919) is a dopamine agonist of the non-ergoline class indicated for treating Parkinson s disease (PD) and restless legs syndrome (RLS) with Ki values of 3 9 nM 2 2 nM 0 5 nM and 5 1 nM for D2S receptor D2L receptor D3 receptor and D4 receptor
Catalog No. 50-311-2222 Supplier Selleck Chemical LLC Supplier No. S506625MG
Pramipexole dihydrochloride (SND919) is a dopamine agonist of the non-ergoline class indicated for treating Parkinson s disease (PD) and restless legs syndrome (RLS) with Ki values of 3 9 nM 2 2 nM 0 5 nM and 5 1 nM for D2S receptor D2L receptor D3 receptor and D4 receptor