Learn More
TARGETMOL CHEMICALS INC Pramipexole dihydrochloride hy
SDP
Catalog No. 502878098
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Pramipexole dihydrochloride hydrate (Mirapex) is the hydrochloride salt of pramipexole, a benzothiazole derivative. As a nonergot dopamine agonist, pramipexole binds to D2 and D3 dopamine receptors in the striatum and substantia nigra of the brain. Purity 99.55%
Catalog No. 50-287-8098 Supplier TARGETMOL CHEMICALS INC Supplier No. T695150MG
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Pramipexole dihydrochloride hydrate (Mirapex) is the hydrochloride salt of pramipexole, a benzothiazole derivative. As a nonergot dopamine agonist, pramipexole binds to D2 and D3 dopamine receptors in the striatum and substantia nigra of the brain. Purity 99.55%