Learn More
Medchemexpress LLC Pramipexole dihydrochloride hydrate | 191217-81-9 | 98.0% | 50 MG
SDP
Catalog No. 5000408080
Encompass_Preferred
Pramipexole dihydrochloride hydrate is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist. It is intended for research in conditions such as Parkinson's disease (PD) and restless legs syndrome (RLS).
  • Selective and blood-brain barrier penetrant dopamine D2-type receptor agonist
  • Ki values for D2-type receptor, D2, D3, and D4 receptors are 2.2 nM, 3.9 nM, 0.5 nM, and 1.3 nM respectively
  • Suitable for Parkinson's disease and restless legs syndrome research
  • Produces dose-dependent increases in dendritic arborization and soma size
  • Attenuates levodopa-induced toxicity in mesencephalic cultures
  • Reduces infarction volume and improves neurological recovery in animal models
Catalog No. 50-004-08080 Supplier Medchemexpress LLC Supplier No. HYB0410A50MG
Pramipexole dihydrochloride hydrate is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist. It is intended for research in conditions such as Parkinson's disease (PD) and restless legs syndrome (RLS).
  • Selective and blood-brain barrier penetrant dopamine D2-type receptor agonist
  • Ki values for D2-type receptor, D2, D3, and D4 receptors are 2.2 nM, 3.9 nM, 0.5 nM, and 1.3 nM respectively
  • Suitable for Parkinson's disease and restless legs syndrome research
  • Produces dose-dependent increases in dendritic arborization and soma size
  • Attenuates levodopa-induced toxicity in mesencephalic cultures
  • Reduces infarction volume and improves neurological recovery in animal models