Learn More
Apexbio Technology LLC Pramiracetam 68497-62-1 200mg
SDP
Catalog No. 5000088735
Encompass_Preferred
Pramiracetam (CAS 68497-62-1) is a small-molecule inhibitor targeting prolyl endopeptidase It is designed to modulate this enzyme thereby influencing neurochemical processes involved in cognitive function Pramiracetam exerts its biological activity primarily through inhibition of prolyl endopeptidase In animal studies pramiracetam demonstrates the ability to modulate cortical nitric oxide synthase (NOS) activity particularly with lithium chloride pretreatment Based on these pharmacological properties pramiracetam holds research potential in the investigation of cognitive impairment following brain injury and in studies of neurodegenerative diseases
Catalog No. 50-000-88735 Supplier Apexbio Technology LLC Supplier No. B2118200
Pramiracetam (CAS 68497-62-1) is a small-molecule inhibitor targeting prolyl endopeptidase It is designed to modulate this enzyme thereby influencing neurochemical processes involved in cognitive function Pramiracetam exerts its biological activity primarily through inhibition of prolyl endopeptidase In animal studies pramiracetam demonstrates the ability to modulate cortical nitric oxide synthase (NOS) activity particularly with lithium chloride pretreatment Based on these pharmacological properties pramiracetam holds research potential in the investigation of cognitive impairment following brain injury and in studies of neurodegenerative diseases