Learn More
Carbosynth LLC. Pramlintide, 5mg, Disulfide bonds: Between Cys2 and Cys7, MW: 3949.47, Molecular formula: C171H267N51O53S2, CAS# 151126-32-8
SDP
Supplier:  Carbosynth LLC. PAM3824PI5MG
Encompass_Preferred
Synonyms: H-Lys-(Cys-Asn-Thr-Ala-Thr-Cys)-Ala-Thr-Gln-Arg-Leu-Ala-Asn-Phe-Leu-Val-His-Ser-Ser-Asn-Asn-Phe-Gly-Pro-Ile-Leu-Pro-Pro-Thr-Asn-Val-Gly-Ser-Asn-Thr-Tyr-NH2; [Pro25,28,29]-Amylin (Human, 1-37); Triproamylin, Symlin, Application: Synthetic analog for amylin pancreatic peptide that has been used for the treatment of Type I and Type II Diabetes as a tool for glycemic control, stimulating glucose production and reducing appetite., Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20°C
Catalog No. 50-168-6121