Learn More
Medchemexpress LLC Pramlintide acetate | 187887-46-3 | 99.89% | 3949.39 (free base) | 100 MG
SDP
Catalog No. 5000425226
Encompass_Preferred
Pramlintide acetate is a synthetic polypeptide analog of human amylin, a neuroendocrine hormone that is co-secreted with insulin from pancreatic β-cells. It functions as an antidiabetic agent and has also shown antineoplastic properties in colorectal cancer.
  • It is a synthetic version of human amylin.
  • Used in the management of diabetes.
  • Demonstrated efficacy against colorectal cancer.
  • Inhibits the growth of HCT-116 and HT-29 cells in a dose-dependent manner.
  • Shows synergistic antiproliferative effects when combined with 5-fluorouracil, Oxaliplatin, or Irinotecan in HCT-116 and HT-29 cells.
  • Intended solely for research purposes.
Catalog No. 50-004-25226 Supplier Medchemexpress LLC Supplier No. HYP0058B100MG
Pramlintide acetate is a synthetic polypeptide analog of human amylin, a neuroendocrine hormone that is co-secreted with insulin from pancreatic β-cells. It functions as an antidiabetic agent and has also shown antineoplastic properties in colorectal cancer.
  • It is a synthetic version of human amylin.
  • Used in the management of diabetes.
  • Demonstrated efficacy against colorectal cancer.
  • Inhibits the growth of HCT-116 and HT-29 cells in a dose-dependent manner.
  • Shows synergistic antiproliferative effects when combined with 5-fluorouracil, Oxaliplatin, or Irinotecan in HCT-116 and HT-29 cells.
  • Intended solely for research purposes.