Learn More
Medchemexpress LLC Pramlintide acetate | 187887-46-3 | 99.9% | 3949.39 | 10 MG
SDP
Catalog No. 5000425531
Encompass_Preferred
Pramlintide acetate is a polypeptide analogue of human amylin. It is an antidiabetic agent and has antineoplastic properties in colorectal cancer.
  • Inhibits the growth of HCT-116 and HT-29 cells in a dose-dependent manner.
  • Synergizes with 5-fluorouracil, Oxaliplatin, or Irinotecan to induce antiproliferative effects in HCT-116 and HT-29 cells.
Catalog No. 50-004-25531 Supplier Medchemexpress LLC Supplier No. HYP0058B10MG
Pramlintide acetate is a polypeptide analogue of human amylin. It is an antidiabetic agent and has antineoplastic properties in colorectal cancer.
  • Inhibits the growth of HCT-116 and HT-29 cells in a dose-dependent manner.
  • Synergizes with 5-fluorouracil, Oxaliplatin, or Irinotecan to induce antiproliferative effects in HCT-116 and HT-29 cells.