Learn More
Medchemexpress LLC Pramlintide acetate | 187887-46-3 | 99.9% | 3949.39 (free base) | 25 MG
SDP
Catalog No. 5000425834
Encompass_Preferred
Pramlintide acetate is a polypeptide analogue of human amylin. It acts as an antidiabetic agent and exhibits antineoplastic activity in colorectal cancer.
  • Inhibits the growth of HCT-116 and HT-29 cells in a dose-dependent manner
  • Demonstrates synergistic antiproliferative effects when combined with 5-fluorouracil, Oxaliplatin, or Irinotecan
Catalog No. 50-004-25834 Supplier Medchemexpress LLC Supplier No. 25MGHYP0058B
Pramlintide acetate is a polypeptide analogue of human amylin. It acts as an antidiabetic agent and exhibits antineoplastic activity in colorectal cancer.
  • Inhibits the growth of HCT-116 and HT-29 cells in a dose-dependent manner
  • Demonstrates synergistic antiproliferative effects when combined with 5-fluorouracil, Oxaliplatin, or Irinotecan