Learn More
Medchemexpress LLC HY-P0058B 5mg , Pramlintide (acetate) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0058B5MG
Encompass_Preferred
HY-P0058B 5mg Pramlintide (acetate) CAS: Pramlintide acetate is a polypeptide analogue of human amylin. Pramlintide acetate, an antidiabetic agent, is antineoplastic in colorectal cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1978