Learn More
Cayman Chemical Pramlintide (acetate hydrate), Purity: ≥95%, CAS: 196078-30-5, 10 mg
SDP
Catalog No. 502934518
Encompass_Preferred
An amylin analog; stimulates cAMP production in HEK293 cells expressing human AMY1a, AMY2a, and AMY3a (EC50s = 0.35, 22.9, and 0.89 nM, respectively); inhibits human islet amyloid polypeptide fibrilization in a concentration-dependent manner; reduces brain levels of Aβ40 and increases spontaneous alternation in the Y-maze in the Tg2576 transgenic mouse model of Alzheimer's disease at 200 pg/kg
Catalog No. 50-293-4518 Supplier Cayman Chemical Supplier No. 2409310 MG
An amylin analog; stimulates cAMP production in HEK293 cells expressing human AMY1a, AMY2a, and AMY3a (EC50s = 0.35, 22.9, and 0.89 nM, respectively); inhibits human islet amyloid polypeptide fibrilization in a concentration-dependent manner; reduces brain levels of Aβ40 and increases spontaneous alternation in the Y-maze in the Tg2576 transgenic mouse model of Alzheimer's disease at 200 pg/kg