Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pramocaine hydrochloride | 637-58-1 | 99.7% | 1 ML
SDP

Catalog No. 5000413213
Encompass_Preferred

Pramocaine hydrochloride decreases the permeability of neuronal membranes to sodium ions, blocking both initiation and conduction of nerve impulses. It is for research use only and not sold to patients.

  • Molecular weight: 329.86
  • Chemical formula: C17H28ClNO3
  • Appearance: solid
  • Color: white to off-white
  • Purity: 99.74%
  • CAS number: 637-58-1
  • Shipping: room temperature in continental US
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-13213 Supplier Medchemexpress LLC Supplier No. HYB131910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pramocaine hydrochloride decreases the permeability of neuronal membranes to sodium ions, blocking both initiation and conduction of nerve impulses. It is for research use only and not sold to patients.

  • Molecular weight: 329.86
  • Chemical formula: C17H28ClNO3
  • Appearance: solid
  • Color: white to off-white
  • Purity: 99.74%
  • CAS number: 637-58-1
  • Shipping: room temperature in continental US
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.