Learn More
Medchemexpress LLC Pramocaine hydrochloride | 637-58-1 | 99.74% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1319500MG
Encompass_Preferred
Pramocaine hydrochloride is an antipruritic agent that decreases the permeability of neuronal membranes to sodium ions, thereby blocking both the initiation and conduction of nerve impulses. This product is for research use only and is not sold to patients.
  • Decreases the permeability of neuronal membranes to sodium ions, blocking the initiation and conduction of nerve impulses.
  • Available with a purity of 99.74%.
  • Offers solubility in both DMSO and H2O, with detailed preparation instructions for stock solutions.
  • Provides specific protocols for in vivo dissolution using various co-solvents such as DMSO, PEG300, Tween-80, Saline, SBE-β-CD, and Corn Oil.
  • Primarily intended as a chemical reagent for research purposes.
  • Reference compound.
Catalog No. 50-004-12492