Learn More
Apexbio Technology LLC Pramoxine HCl 637-58-1 1g
SDP
Catalog No. 5000094301
Encompass_Preferred
Pramoxine HCl is a local anesthetic that transiently inhibits sensory nerve signal conduction through sodium channel blockade in neuronal membranes thereby reducing local sensory nerve impulses and producing anesthetic and antipruritic effects Pramoxine HCl exerts its biological activity primarily through sodium channel blockade Based on these pharmacological properties Pramoxine HCl holds research potential in the investigation of sensory nerve inhibition mechanisms pruritus management and in vitro nerve conduction assays
Catalog No. 50-000-94301 Supplier Apexbio Technology LLC Supplier No. B18191000
Add to Cart
Add to Cart
Pramoxine HCl is a local anesthetic that transiently inhibits sensory nerve signal conduction through sodium channel blockade in neuronal membranes thereby reducing local sensory nerve impulses and producing anesthetic and antipruritic effects Pramoxine HCl exerts its biological activity primarily through sodium channel blockade Based on these pharmacological properties Pramoxine HCl holds research potential in the investigation of sensory nerve inhibition mechanisms pruritus management and in vitro nerve conduction assays