Learn More
Cayman Chemical Pramoxine (hydrochloride), Purity: ≥98%, CAS: 637-58-1, 50 g
SDP
Catalog No. 502990375
Encompass_Preferred
A local anesthetic; reduces electrically induced peak CAP amplitudes in isolated frog sciatic nerve (IC50 = 0.21 mM); topical administration inhibits winking in a rabbit corneal test, indicating anesthetic activity, at 0.35 and 0.6%; inhibits the pinprick-induced CTMR in the skin of rats (ED50 = 42.1 μmol, s.c.)
Catalog No. 50-299-0375 Supplier Cayman Chemical Supplier No. 3007750 G
A local anesthetic; reduces electrically induced peak CAP amplitudes in isolated frog sciatic nerve (IC50 = 0.21 mM); topical administration inhibits winking in a rabbit corneal test, indicating anesthetic activity, at 0.35 and 0.6%; inhibits the pinprick-induced CTMR in the skin of rats (ED50 = 42.1 μmol, s.c.)