Learn More
Medchemexpress LLC Pranidipine (OPC-13340) | 99522-79-9 | 99.5% | 448.47 | 50 MG
SDP
Catalog No. 5000400938
Encompass_Preferred
Pranidipine (OPC-13340) is a potent, long-acting 1,4-dihydropyridine calcium channel blocker with antihypertensive activity. It is for research use only.
  • Potent, long-acting 1,4-dihydropyridine calcium channel blocker.
  • Antihypertensive activity.
  • For research use only.
  • High purity.
Catalog No. 50-004-00938 Supplier Medchemexpress LLC Supplier No. HY1966450MG
Pranidipine (OPC-13340) is a potent, long-acting 1,4-dihydropyridine calcium channel blocker with antihypertensive activity. It is for research use only.
  • Potent, long-acting 1,4-dihydropyridine calcium channel blocker.
  • Antihypertensive activity.
  • For research use only.
  • High purity.