Learn More
Medchemexpress LLC HY-B0290 100mg Medchemexpress, Pranlukast CAS:103177-37-3 Purity:>98%
SDP
Catalog No. 502082450
Encompass_Preferred
Medchemexpress, HY-B0290 100mg Pranlukast CAS:103177-37-3 Pranlukast is a highly potent, selective and competitive antagonist of peptide leukotrienes. Pranlukast inhibits [3H]LTE4, [3H]LTD4, and [3H]LTC4 bindings to lung membranes with Kis of 0.63±0.11, 0.99±0.19, and 5640±680 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2450 Supplier Medchemexpress LLC Supplier No. HYB0290100MG
Medchemexpress, HY-B0290 100mg Pranlukast CAS:103177-37-3 Pranlukast is a highly potent, selective and competitive antagonist of peptide leukotrienes. Pranlukast inhibits [3H]LTE4, [3H]LTD4, and [3H]LTC4 bindings to lung membranes with Kis of 0.63±0.11, 0.99±0.19, and 5640±680 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.