Learn More
Apexbio Technology LLC Pranlukast 103177-37-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061993
Encompass_Preferred
Pranlukast (CAS 103177-37-3) is a selective antagonist of cysteinyl leukotriene (cysLT) receptors exhibiting its pharmacological action by competitively binding to targeted leukotriene receptor sites It specifically prevents the interaction of endogenous leukotrienes (LTC4 LTD4 LTE4) with corresponding receptors located on pulmonary cell membranes thus reducing leukotriene-driven airway inflammation and bronchoconstriction Binding experiments demonstrate that pranlukast possesses notably greater affinity for LTE4 and LTD4 compared to LTC4 In biomedical research pranlukast is frequently utilized for mechanistic studies and pharmacological modeling of asthma allergic rhinitis and related airway inflammatory conditions
Catalog No. 50-000-61993 Supplier Apexbio Technology LLC Supplier No. B20095.1
Pranlukast (CAS 103177-37-3) is a selective antagonist of cysteinyl leukotriene (cysLT) receptors exhibiting its pharmacological action by competitively binding to targeted leukotriene receptor sites It specifically prevents the interaction of endogenous leukotrienes (LTC4 LTD4 LTE4) with corresponding receptors located on pulmonary cell membranes thus reducing leukotriene-driven airway inflammation and bronchoconstriction Binding experiments demonstrate that pranlukast possesses notably greater affinity for LTE4 and LTD4 compared to LTC4 In biomedical research pranlukast is frequently utilized for mechanistic studies and pharmacological modeling of asthma allergic rhinitis and related airway inflammatory conditions