Learn More
TARGETMOL CHEMICALS INC Pranlukast 200MG
SDP
Catalog No. 502880722
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Pranlukast (ONO-1078) is a cysteinyl leukotriene receptor-1 antagonist. It antagonizes or reduces bronchospasm caused, principally in asthmatics, by an allergic reaction to accidentally or inadvertently encountered allergens. Purity 99.85%
Catalog No. 50-288-0722 Supplier TARGETMOL CHEMICALS INC Supplier No. T0694200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Pranlukast (ONO-1078) is a cysteinyl leukotriene receptor-1 antagonist. It antagonizes or reduces bronchospasm caused, principally in asthmatics, by an allergic reaction to accidentally or inadvertently encountered allergens. Purity 99.85%