Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pranlukast hemihydrate | 150821-03-7 | MFCD24539454 | 99.3% | 50 MG
SDP

Catalog No. 5000408992
Encompass_Preferred

Pranlukast hemihydrate is a highly potent, selective, and competitive antagonist of peptide leukotrienes, acting as a subtype-specific CysLT1 receptor antagonist. Leukotrienes are involved in the inflammation response.

  • Highly potent, selective, and competitive antagonist of peptide leukotrienes.
  • Inhibits [3H]LTE4, [3H]LTD4, and [3H]LTC4 bindings to lung membranes.
  • Ki for LTE4 is 0.63 nM.
  • Ki for LTD4 is 0.99 nM.
  • Ki for LTC4 is 5640 nM.

Catalog No. 50-004-08992 Supplier Medchemexpress LLC Supplier No. HYB0290A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pranlukast hemihydrate is a highly potent, selective, and competitive antagonist of peptide leukotrienes, acting as a subtype-specific CysLT1 receptor antagonist. Leukotrienes are involved in the inflammation response.

  • Highly potent, selective, and competitive antagonist of peptide leukotrienes.
  • Inhibits [3H]LTE4, [3H]LTD4, and [3H]LTC4 bindings to lung membranes.
  • Ki for LTE4 is 0.63 nM.
  • Ki for LTD4 is 0.99 nM.
  • Ki for LTC4 is 5640 nM.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.