Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Pranlukast hemihydrate

Catalog No. AC465335000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465335000 500 mg
AC465331000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465335000 Supplier Thermo Scientific Chemicals Supplier No. 465335000
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150821-03-7
Molecular Formula C54H48N10O9
Molecular Weight (g/mol) 981.04
InChI Key MSXTUBJFNBZPGC-UHFFFAOYSA-N
IUPAC Name bis(N-[4-oxo-2-(2H-1,2,3,4-tetrazol-5-yl)-4H-chromen-8-yl]-4-(4-phenylbutoxy)benzamide) hydrate
SMILES O.O=C(NC1=C2OC(=CC(=O)C2=CC=C1)C1=NNN=N1)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1.O=C(NC1=C2OC(=CC(=O)C2=CC=C1)C1=NNN=N1)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1

Specifications

Quantity 500 mg
Formula Weight 490.51
Chemical Name or Material Pranlukast hemihydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

May cause long lasting harmful effects to aquatic life.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.