Learn More
TARGETMOL CHEMICALS INC Pranoprofen 200MG
SDP
Catalog No. 502880546
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Pranoprofen (Pyranoprofen) (INN) is a non-steroidal anti-inflammatory drug used in ophthalmology. Purity 97.99%
Catalog No. 50-288-0546 Supplier TARGETMOL CHEMICALS INC Supplier No. T0159200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Pranoprofen (Pyranoprofen) (INN) is a non-steroidal anti-inflammatory drug used in ophthalmology. Purity 97.99%