Learn More
Aviva Systems Biology Corp PRAP1 Antibody - C-terminal region (ARP58517_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58517_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRAP1. It was validated on Western Blot. . Alternate names: MGC126792, PRO1195, RP11-122K13.6, UPAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-286-480