Learn More
Biolog Inc PRAS Dilution Blanks, 9 mL, 10 Tubes/pack
SDP
Catalog No. NC1701792
Encompass
This item has a minimum qty of 3 per supplier requirements.
Dilution Blanks are a buffered mineral salt base solution with reducing agents, designed as a holding medium for maintaining viability of microorganisms, especially anaerobes, in order to determine the number of viable organisms present in a sample.
Catalog No. NC1701792 Supplier Biolog Inc Supplier No. AS908
This item has a minimum qty of 3 per supplier requirements.
Dilution Blanks are a buffered mineral salt base solution with reducing agents, designed as a holding medium for maintaining viability of microorganisms, especially anaerobes, in order to determine the number of viable organisms present in a sample.